missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human ACVR2B Protein

A DNA sequence encoding the Homo sapiens (Human) ACVR2B, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9768-ye-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16028691

  • € 2765.00 / 1mg
Estimated Shipment: 21-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001097.2
ACVR2B Protein
Research Use Only
ACVR2B
Recombinant Protein
Yeast
SGRGEAETRE CIYYNANWEL ERTNQSGLER CEGEQDKRLH CYASWRNSSG TIELVKKGCW LDDFNCYDRQ ECVATEENPQ VYFCCCEGNF CNERFTHLPE AGGPEVTYEP PPTAPTLLT
Greater than 90% as determined by SDS-PAGE.
93
1 mg
Not determined.
Extracellular Domain
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers