missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human ADH7 Protein

A DNA sequence encoding the Homo sapiens (Human) ADH7, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP5617-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15936469

  • € 458.00 / 100µg

Estimated Shipment: 30-01-2023
to see stock.
Add to basket

Description

Description

Specifications

Specifications

NP_000664.2
ADH7 Protein
100 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
131
Greater than 90% as determined by SDS-PAGE.
Research Use Only
ADH7
Recombinant Protein
E. coli
MFAEIQIQDK DRMGTAGKVI KCKAAVLWEQ KQPFSIEEIE VAPPKTKEVR IKILATGICR TDDHVIKGTM VSKFPVIVGH EATGIVESIG EGVTTVKPGD KVIPLFLPQC RECNACRNPD GNLCIRSDIT GRGVLADGTT RFTCKGKPVH HFMNTSTFTE YTVVDESSVA KIDDAAPPEK VCLIGCGFST GYGAAVKTGK VKPGSTCVVF GLGGVGLSVI MGCKSAGASR IIGIDLNKDK FEKAMAVGAT ECISPKDSTK PISEVLSEMT GNNVGYTFEV IGHLETMIDA LASCHMNYGT SVVVGVPPSA KMLTYDPMLL FTGRTWKGCV FGGLKSRDDV PKLVTEFLAK KFDLDQLITH VLPFKKISEG FELLNSGQSI RTVLTF
Videos
SDS
Documents
Certificates
Special Offers

Special Offers