missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human Aggrecan G1-IGD-G2 Domains Protein

Product Code. 18147690
Click to view available options
Quantity:
25 μg
Unit Size:
25µg
This item is not returnable. View return policy

Product Code. 18147690

Brand: R&D Systems™ 1220PG025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Enzyme Activity, Binding Activity

The Recombinant Human Aggrecan G1-IGD-G2 Domains Protein is derived from NS0. The Recombinant Human Aggrecan G1-IGD-G2 Domains Protein has been validated for the following applications: Enzyme Activity, Binding Activity.
TRUSTED_SUSTAINABILITY

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in Tris and NaCl.
Gene ID (Entrez) 176
Molecular Weight (g/mol) 74 kDa
Name Aggrecan G1-IGD-G2 Domains
Quantity 25 μg
Source Mouse myeloma cell line,NS0-derived human Aggrecan protein Val20-Gly675,with a C-terminal 10-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution For hyaluronan-binding assay, reconstitute at 100 μg/mL in PBS. For use as a metalloprotease substrate, reconstitute at 250 μg/mL in 50 mM Tris, 10 mM CaCl 2 150 mM NaCl and 0.05% Brij-35, pH 7.5.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie Blue Staining.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.