missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human ALAD Protein

A cDNA sequence encoding the ALAD was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP10996-2ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15935818

  • € 137.00 / 2µg
Estimated Shipment: 26-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

210
2 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSHMQPQSV LHSGYFHPLL RAWQTATTTL NASNLIYPIF VTDVPDDIQP ITSLPGVARY GVKRLEEMLR PLVEEGLRCV LIFGVPSRVP KDERGSAADS EESPAIEAIH LLRKTFPNLL VACDVCLCPY TSHGHCGLLS ENGAFRAEES RQRLAEVALA YAKAGCQVVA PSDMMDGRVE AIKEALMAHG LGNRVSVMSY SAKFASCFYG PFRDAAKSSP AFGDRRCYQL PPGARGLALR AVDRDVREGA DMLMVKPGMP YLDIVREVKD KHPDLPLAVY HVSGEFAMLW HGAQAGAFDL KAAVLEAMTA FRRAGADIII TYYTPQLLQW LKEE
Greater than 85% as determined by SDS-PAGE.
ALAD Protein
Research Use Only
ALAD
Human
His
The ALAD solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 100mM NaCl and 10% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers