missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Alanine aminotransferase 1 Protein

A DNA sequence encoding the Homo sapiens (Human) Alanine aminotransferase 1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6114-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16030900

  • € 530.00 / 100µg
Estimated Shipment: 29-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_005300.1
Alanine aminotransferase 1 Protein
Research Use Only
GPT
Recombinant Protein
E. coli
ASSTGDRSQA VRHGLRAKVL TLDGMNPRVR RVEYAVRGPI VQRALELEQE LRQGVKKPFT EVIRANIGDA QAMGQRPITF LRQVLALCVN PDLLSSPNFP DDAKKRAERI LQACGGHSLG AYSVSSGIQL IREDVARYIE RRDGGIPADP NNVFLSTGAS DAIVTVLKLL VAGEGHTRTG VLIPIPQYPL YSATLAELGA VQVDYYLDEE RAWALDVAEL HRALGQARDH CRPRALCVIN PGNPTGQVQT RECIEAVIRF AFEERLFLLA DEVYQDNVYA AGSQFHSFKK VLMEMGPPYA GQQELASFHS TSKGYMGECG FRGGYVEVVN MDAAVQQQML KLMSVRLCPP VPGQALLDLV VSPPAPTDPS FAQFQAEKQA VLAELAAKAK LTEQVFNEAP GISCNPVQGA MYSFPRVQLP PRAVERAQEL GLAPDMFFCL RLLEETGICV VPGSGFGQRE GTYHFRMTIL PPLEKLRLLL EKLSRFHAKF TLEYS
Greater than 90% as determined by SDS-PAGE.
2875
100 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers