missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human alpha 2-Macroglobulin-like 1 Protein
Click to view available options
:
50ug; Carrier Free
Description
The Recombinant Human alpha 2-Macroglobulin-like 1 Protein is derived from HEK293. The Recombinant Human alpha 2-Macroglobulin-like 1 Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS. |
| Gene ID (Entrez) | 144568 |
| Molecular Weight (g/mol) | 160 kDa |
| Name | alpha 2-Macroglobulin-like 1/A2ML1 |
| Quantity | 50μg |
| Source | Human embryonic kidney cell,HEK293-derived human alpha 2-Macroglobulin-like 1/A2ML1 protein Glu18-Glu1454,with a C-terminal 6-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Purity or Quality Grade | 75%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction