missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human alpha-L-Iduronidase/IDUA His Protein
Click to view available options
Quantity:
20 μg
Unit Size:
20µg
Description
Measured by its ability to cleave a fluorogenic substrate, 4-Methylumbelliferyl alpha-L-iduronide. The specific activity is >6000 pmol/min/μg, as measured under the described conditions.
Specifications
Specifications
| Accession Number | P35475.2 |
| For Use With (Application) | Enzyme Activity |
| Formulation | Supplied as a 0.2 μm filtered solution in Sodium Acetate, NaCl and Glycerol. |
| Gene ID (Entrez) | 3425 |
| Molecular Weight (g/mol) | M.W. Observed: 80-91 kDa, under reducing conditions., M.W. Theoretical: 71 kDa |
| Quantity | 20 μg |
| Source | Chinese Hamster Ovary cell line, CHO-derived human alpha-L-Iduronidase/IDUA protein Ala28-Pro653 (His33Gln), with a C-terminal 10-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70 °C as supplied. 3 months, -20 to -70 °C under sterile conditions after opening. |
| Endotoxin Concentration | <0.10 EU per 1 μg of the protein by the LAL method. |
| Gene Alias | alphaLIduronidase, IDA, IDUA, MPS1, MPSI |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction