missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Alpha-L-iduronidase Protein
A DNA sequence encoding the Homo sapiens (Human) Alpha-L-iduronidase, was expressed in the hosts and tags indicated.
€ 202.00 - € 2155.00
Specifications
Accession Number | NP_000194.2 |
---|---|
Gene ID (Entrez) | 3425 |
Name | Alpha-L-iduronidase Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16031500
|
enQuireBio™
QP6197-YE-100UG |
100 μg |
€ 645.00
100µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16071500
|
enQuireBio™
QP6197-YE-500UG |
500 μg |
€ 1740.00
500µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16041500
|
enQuireBio™
QP6197-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16061500
|
enQuireBio™
QP6197-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16081500
|
enQuireBio™
QP6197-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16051500
|
enQuireBio™
QP6197-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_000194.2 | |
Alpha-L-iduronidase Protein | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
3425 | |
Research Use Only | |
IDUA | |
Recombinant Protein | |
Yeast | |
APHLVHVDAA RALWPLRRFW RSTGFCPPLP HSQADQYVLS WDQQLNLAYV GAVPHRGIKQ VRTHWLLELV TTRGSTGRGL SYNFTHLDGY LDLLRENQLL PGFELMGSAS GHFTDFEDKQ QVFEWKDLVS SLARRYIGRY GLAHVSKWNF ETWNEPDHHD FDNVSMTMQG FLNYYDACSE GLRAASPALR LGGPGDSFHT PPRSPLSWGL LRHCHDGTNF FTGEAGVRLD YISLHRKGAR SSISILEQEK VVAQQIRQLF PKFADTPIYN DEADPLVGWS LPQPWRADVT YAAMVVKVIA QHQNLLLANT TSAFPYALLS NDNAFLSYHP HPFAQRTLTA RFQVNNTRPP HVQLLRKPVL TAMGLLALLD EEQLWAEVSQ AGTVLDSNHT VGVLASAHRP QGPADAWRAA VLIYASDDTR AHPNRSVAVT LRLRGVPPGP GLVYVTRYLD NGLCSPDGEW RRLGRPVFPT AEQFRRMRAA EDPVAAAPRP LPAGGRLTLR PALRLPSLLL VHVCARPEKP PGQVTRLRAL PLTQGQLVLV WSDEHVGSKC LWTYEIQFSQ DGKAYTPVSR KPSTFNLFVF SPDTGAVSGS YRVRALDYWA RPGPFSDPVP YLEVPVPRGP PSPGNP | |
Greater than 90% as determined by SDS-PAGE. |