missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Alpha-L-iduronidase Protein

A DNA sequence encoding the Homo sapiens (Human) Alpha-L-iduronidase, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6197-ye-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16031500

  • € 491.00 / 100µg


Estimated Shipment
29-10-2020
Add to basket

Specifications

Specifications

NP_000194.2
Alpha-L-iduronidase Protein
100 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
3425
Greater than 90% as determined by SDS-PAGE.
Research Use Only
IDUA
Recombinant Protein
Yeast
APHLVHVDAA RALWPLRRFW RSTGFCPPLP HSQADQYVLS WDQQLNLAYV GAVPHRGIKQ VRTHWLLELV TTRGSTGRGL SYNFTHLDGY LDLLRENQLL PGFELMGSAS GHFTDFEDKQ QVFEWKDLVS SLARRYIGRY GLAHVSKWNF ETWNEPDHHD FDNVSMTMQG FLNYYDACSE GLRAASPALR LGGPGDSFHT PPRSPLSWGL LRHCHDGTNF FTGEAGVRLD YISLHRKGAR SSISILEQEK VVAQQIRQLF PKFADTPIYN DEADPLVGWS LPQPWRADVT YAAMVVKVIA QHQNLLLANT TSAFPYALLS NDNAFLSYHP HPFAQRTLTA RFQVNNTRPP HVQLLRKPVL TAMGLLALLD EEQLWAEVSQ AGTVLDSNHT VGVLASAHRP QGPADAWRAA VLIYASDDTR AHPNRSVAVT LRLRGVPPGP GLVYVTRYLD NGLCSPDGEW RRLGRPVFPT AEQFRRMRAA EDPVAAAPRP LPAGGRLTLR PALRLPSLLL VHVCARPEKP PGQVTRLRAL PLTQGQLVLV WSDEHVGSKC LWTYEIQFSQ DGKAYTPVSR KPSTFNLFVF SPDTGAVSGS YRVRALDYWA RPGPFSDPVP YLEVPVPRGP PSPGNP
Videos
Documents