missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human AMBP Protein

A cDNA sequence encoding the AMBP was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11009-HIS-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15935858

  • € 7200.00 / 1mg
Estimated Shipment: 22-11-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

AMBP Protein
Research Use Only
Recombinant Protein
E. coli
MRSLGALLLL LSACLAVSAG PVPTPPDNIQ VQENFNISRI YGKWYNLAIG STCPWLKKIM DRMTVSTLVL GEGATEAEIS MTSTRWRKGV CEETSGAYEK TDTDGKFLYH KSKWNITMES YVVHTNYDEY AIFLTKKFSR HHGPTITAKL YGRAPQLRET LLQDFRVVAQ GVGIPEDSIF TMADRGECVP GEQEPEPILI PRVRRAVLPQ EEEGSGGGQL VTEVTKKEDS CQLGYSAGPC MGMTSRYFYN GTSMACETFQ YGGCMGNGNN FVTEKECLQT CRTVAACNLP IVRGPCRAFI QLWAFDAVKG KCVLFPYGGC QGNGNKFYSE KECREYCGVP GDGDEELLRF SN
Greater than 96.0%.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
Lyophilized from 0.02M NH4HCO3.
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers