missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human AMMECR1L Protein
A cDNA sequence encoding the AMMECR1L was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11012-5ug
Additional Details : Weight : 0.01000kg
Specifications
AMMECR1L Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMGKRRCV PPLEPKLAAG CCGVKKPKLS GSGTHSHGNQ STTVPGSSSG PLQNHQHVDS SSGRENVSDL TLGPGNSPIT RMNPASGALS PLPRPNGTAN TTKNLVVTAE MCCYCFDVLY CHLYGFPQPR LPRFTNDPYP LFVTWKTGRD KRLRGCIGTF SAMNLHSGLR EYTLTSALKD SRFPPLTREE LPKLFCSVSL LTNFEDASDY LDWEVGVHGI RIEFINEKGV KRTATYLPEV AKEQDWDQIQ TIDSLLRKGG FKAPITSEFR KTIKLTRYRS EKVTISYAEY IASRQHCFQN GTLHAPPLYN HYS | |
Greater than 90% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The AMMECR1L solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 10% glycerol and 1mM DTT. |