missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Annexin A8-like protein 1
A DNA sequence encoding the Homo sapiens (Human) Annexin A8-like protein 1, was expressed in the hosts and tags indicated.
Marke: enQuireBio™ QP7711-ec-10ug
Additional Details : Weight : 0.01000kg
Spezifikation
NP_001092315.2 | |
Annexin A8-like protein 1 | |
Research Use Only | |
ANXA8L2 | |
Recombinant Protein | |
E. coli | |
MAWWKAWIEQ EGVTVKSSSH FNPDPDAETL YKAMKGIGVG SQLLSHQAAA FAFPSSALTS VSPWGQQGHL CCNPAGTNEQ AIIDVLTKRS NTQRQQIAKS FKAQFGKDLT ETLKSELSGK FERLIVALMY PPYRYEAKEL HDAMKGSRDD VSSFVDPALA LQDAQDLYAA GEKIRGTDEM KFITILCTRS ATHLLRVKCT QNLHSYFAER LYYAMKGAGT RDGTLIRNIV SRSEIDLNLI KCHFKKMYGK TLSSMIMEDT SGDYKNALLS LVGSDP | |
Greater than 80% as determined by SDS-PAGE. |
728113 | |
10 μg | |
Not determined. | |
Full Length of Isoform 2 | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |