missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human AP1AR Protein
A cDNA sequence encoding the AP1AR was constructed and used to recombinantly synthesize the protein.
€ 137.00 - € 1815.00
Specifications
Gene ID (Entrez) | 55435 |
---|---|
Name | AP1AR Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gene Symbol | AP1AR |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15935948
|
enQuireBio™
QP11038-2UG |
2 μg |
€ 137.00
2µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15925948
|
enQuireBio™
QP11038-10UG |
10 μg |
€ 212.00
10µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15915948
|
enQuireBio™
QP11038-100UG |
100 μg |
€ 1815.00
100µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
55435 | |
Research Use Only | |
AP1AR | |
Human | |
His | |
The AP1AR solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 10% glycerol and 1mM DTT. |
AP1AR Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGNCCWTQCF GLLRKEAGRL QRVGGGGGSK YFRTCSRGEH LTIEFENLVE SDEGESPGSS HRPLTEEEIV DLRERHYDSI AEKQKDLDKK IQKELALQEE KLRLEEEALY AAQREAARAA KQRKLLEQER QRIVQQYHPS NNGEYQSSGP EDDFESCLRN MKSQYEVFRS SRLSSDATVL TPNTESSCDL MTKTKSTSGN DDSTSLDLEW EDEEGMNRML PMRERSKTEE DILRAALKYS NKKTGSNPTS ASDDSNGLEW ENDFVSAEMD DNGNSEYSGF VNPVLELSDS GIRHSDTDQQ TR | |
Greater than 90.0% as determined by SDS-PAGE. |