missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human Apolipoprotein A-IV/ApoA4 Protein, CF

Product Code. 18107480
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
This item is not returnable. View return policy

Product Code. 18107480

Brand: R&D Systems™ 9606AP050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Carrier-Free Recombinant Protein

Bioassay data are not available.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P06727
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS.
Gene ID (Entrez) 337
Molecular Weight (g/mol) M.W. (Observed): 43 kDa, reducing conditions, M.W. (Theoretical): 44 kDa
Quantity 50 μg
Source E. coli-derived human Apolipoprotein A-IV/ApoA4 protein Glu21-Ser396, with an N-terminal Met
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1 μg of the protein by the LAL method.
Gene Alias apo-AIV, ApoA-IV, Apolipoprotein A4, apolipoprotein A-IV, Apolipoprotein AIV, MGC142154, MGC142156
Conjugate Unconjugated
Reconstitution Reconstitute at 250 μg/mL in PBS.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein Apolipoprotein A-IV/ApoA4
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.