missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human Apolipoprotein E3 Protein
Click to view available options
Quantity:
500 μg
Unit Size:
500µg
Description
The Recombinant Human Apolipoprotein E3 Protein is derived from E. coli. The Recombinant Human Apolipoprotein E3 Protein has been validated for the following applications: Binding Activity.
Specifications
Specifications
| Formulation | Supplied as a 0.2μm filtered solution in MOPS, NaCl, CHAPS and TCEP. |
| Gene ID (Entrez) | 348 |
| Molecular Weight (g/mol) | 35.2 kDa |
| Name | Apolipoprotein E3/ApoE3 |
| Quantity | 500 μg |
| Source | E. coli-derived human Apolipoprotein E3/ApoE3 protein Lys19-His317,with an N-terminal Met and 6-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after opening. 3 months, -20 to -70° C under sterile conditions after opening. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Purity or Quality Grade | 95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction