missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Apoptosis-inducing factor 1, mitochondrial Protein
A DNA sequence encoding the Homo sapiens (Human) Apoptosis-inducing factor 1, mitochondrial, was expressed in the hosts and tags indicated.
€ 160.00 - € 2385.00
Specifications
Accession Number | NP_001124318.2 |
---|---|
Gene ID (Entrez) | 9131 |
Name | Apoptosis-inducing factor 1, mitochondrial Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15936589
|
enQuireBio™
QP5633-EC-100UG |
100 μg |
€ 530.00
100µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15956589
|
enQuireBio™
QP5633-EC-1MG |
1 mg |
€ 2385.00
1mg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15946589
|
enQuireBio™
QP5633-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15966589
|
enQuireBio™
QP5633-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15976589
|
enQuireBio™
QP5633-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15986589
|
enQuireBio™
QP5633-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001124318.2 | |
Apoptosis-inducing factor 1, mitochondrial Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
9131 | |
Research Use Only | |
AIFM1 | |
Recombinant Protein | |
E. coli | |
GLTPEQKQKK AALSASEGEE VPQDKAPSHV PFLLIGGGTA AFAAARSIRA RDPGARVLIV SEDPELPYMR PPLSKELWFS DDPNVTKTLR FKQWNGKERS IYFQPPSFYV SAQDLPHIEN GGVAVLTGKK VVQLDVRDNM VKLNDGSQIT YEKCLIATGG TPRSLSAIDR AGAEVKSRTT LFRKIGDFRS LEKISREVKS ITIIGGGFLG SELACALGRK ARALGTEVIQ LFPEKGNMGK ILPEYLSNWT MEKVRREGVK VMPNAIVQSV GVSSGKLLIK LKDGRKVETD HIVAAVGLEP NVELAKTGGL EIDSDFGGFR VNAELQARSN IWVAGDAACF YDIKLGRRRV EHHDHAVVSG RLAGENMTGA AKPYWHQSMF WSDLGPDVGY EAIGLVDSSL PTVGVFAKAT AQDNPKSATE QSGTGIRSES ETESEASEIT IPPSTPAVPQ APVQGEDYGK GVIFYLRDKV VVGIVLWNIF NRMPIARKII KDGEQHEDLN EVAKLFNIHE | |
Greater than 90% as determined by SDS-PAGE. |