missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Atrial natriuretic peptide receptor 1 Protein

Product Code. 16033260
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
This item is not returnable. View return policy

Product Code. 16033260

Brand: enQuireBio™ QP6431ec1mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A DNA sequence encoding the Homo sapiens (Human) Atrial natriuretic peptide receptor 1, was expressed in the hosts and tags indicated.

Specifications

Accession Number NP_000897.3
Gene ID (Entrez) 4881
Name Atrial natriuretic peptide receptor 1 Protein
Quantity 1 mg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol NPR1
Protein Length Extracellular Domain
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag His-SUMO
Sequence GNLTVAVVLP LANTSYPWSW ARVGPAVELA LAQVKARPDL LPGWTVRTVL GSSENALGVC SDTAAPLAAV DLKWEHNPAV FLGPGCVYAA APVGRFTAHW RVPLLTAGAP ALGFGVKDEY ALTTRAGPSY AKLGDFVAAL HRRLGWERQA LMLYAYRPGD EEHCFFLVEG LFMRVRDRLN ITVDHLEFAE DDLSHYTRLL RTMPRKGRVI YICSSPDAFR TLMLLALEAG LCGEDYVFFH LDIFGQSLQG GQGPAPRRPW ERGDGQDVSA RQAFQAAKII TYKDPDNPEY LEFLKQLKHL AYEQFNFTME DGLVNTIPAS FHDGLLLYIQ AVTETLAHGG TVTDGENITQ RMWNRSFQGV TGYLKIDSSG DRETDFSLWD MDPENGAFRV VLNYNGTSQE LVAVSGRKLN WPLGYPPPDI PKCGFDNEDP ACNQDHLSTL E
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.