missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Autophagy-related protein 16-1 Protein

A DNA sequence encoding the Homo sapiens (Human) Autophagy-related protein 16-1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7727-ec-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16070451

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001177195.1
Autophagy-related protein 16-1 Protein
Research Use Only
ATG16L1
Recombinant Protein
E. coli
MSSGLRAADF PRWKRHISEQ LRRRDRLQRQ AFEEIILQYN KLLEKSDLHS VLAQKLQAEK HDVPNRHEIS PGHDGTWNDN QLQEMAQLRI KHQEELTELH KKRGELAQLV IDLNNQMQRK DREMQMNEAK IAECLQTISD LETECLDLRT KLCDLERANQ TLKDEYDALQ ITFTALEGKL RKTTEENQEL VTRWMAEKAQ EANRLNAENE KDSRRRQARL QKELAEAAKE PLPVEQDDDI EVIVDETSDH TEETSPVRAI SRAATKRLSQ PAGGLLDSIT NIFGRRSVSS FPVPQDNVDT HPGSGKEVRV PATALCVFDA HDGEVNAVQF SPGSRLLATG GMDRRVKLWE VFGEKCEFKG SLSGSNAGIT SIEFDSAGSY LLAASNDFAS RIWTVDDYRL RHTLTGHSGK VLSAKFLLDN ARIVSGSHDR TLKLWDLRSK VCIKTVFAGS SCNDIVCTEQ CVMSGHFDKK IRFWDIRSES IVREMELLGK ITALDLNPER TELLSCSRDD LLKVIDLRTN AIKQTFSAPG FKCGSDWTRV VFSPDGSYVA AGSAEGSLYI WSVLTGKVEK VLSKQHSSSI NAVAWSPSGS HVVSVDKGCK AVLWAQY
Greater than 90% as determined by SDS-PAGE.
55054
500 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers