missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human B3GAT3 Protein
Click to view available options
Quantity:
20 μg
Unit Size:
20µg
Description
Measured by its ability to hydrolyze UDP-GlcA. The specific activity is >1 pmol/min/μg, as measured under the described conditions.
Specifications
Specifications
| Accession Number | O94766 |
| For Use With (Application) | Enzyme Activity |
| Formulation | Supplied as a 0.2 μm filtered solution in Tris and NaCl. |
| Gene ID (Entrez) | 26229 |
| Molecular Weight (g/mol) | M.W. (Observed): 30-35 kDa, reducing conditions; M.W. (theoretical): 30 kDa |
| Name | beta-1,3-Glucuronyltransferase 3/B3GAT3 |
| Quantity | 20 μg |
| Source | E. coli-derived human beta-1,3-Glucuronyltransferase 3/B3GAT3 protein Glu72-Val335, with an N-terminal Met and 6-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening. |
| Endotoxin Concentration | <1.0 EU / 1 μg of the protein by the LAL method. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction