missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human B7-2/CD86 Fc Alexa Fluor™ 647 Protein

Product Code. 18718756
Click to view available options
Quantity:
20 μg
Unit Size:
20µg
This item is not returnable. View return policy

Product Code. 18718756

Brand: R&D Systems™ AFR7625020

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Fc Chimera

Measured by flow cytometry for its ability to bind anti-human B7-2/CD86 Antibody conjugated beads. The concentration of Recombinant Human B7-2/CD86 Fc Chimera Alexa Fluor™ 647 (Catalog # AFR7625) that produces 50% of the binding response is 2.50-25.0 ng/mL.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number AAB03814.1
For Use With (Application) Bioactivity
Formulation Supplied as a 0.2 μm filtered solution in PBS with BSA as a carrier protein.
Gene ID (Entrez) 942
Molecular Weight (g/mol) M.W.-observed: 75-100 kDa, under reducing conditions., M.W.-theroretical: 52 kDa (monomer)
Quantity 20 μg
Source Chinese Hamster Ovary cell line, CHO-derived human B7-2/CD86 protein Human B7-2/CD86 (Leu20-His239) (C-terminus) Accession # AAB03814.1 DIEGRMD Human IgG1 (Pro100-Lys330) (N-terminus)
Storage Requirements Protect from light. Use a manual defrost freezer and avoid repeated freeze-thaw cycles.
Endotoxin Concentration <1.0 EU / 1 μg of the protein by the LAL method.
Gene Alias Activation B7-2 antigen, B70, B72, B7-2, B7-2 antigen, B-lymphocyte activation antigen B7-2, BU63, CD28 antigen ligand 2, CD28LG2B7-2 antigen), CD86, CD86 antigen, CD86 molecule, CTLA-4 counter-receptor B7.2, FUN-1, LAB72, MGC34413, T-lymphocyte activation antigen CD86
Conjugate Alexa Fluor 647
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein B7-2/CD86
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.