missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human B7-H2 Fc Chimera Alexa Fluor™ 488 Protein
Click to view available options
Quantity:
20 μg
Unit Size:
20µg
Description
Measured by flow cytometry for its ability to bind anti-human B7-H2 Antibody conjugated beads. The concentration of Recombinant Human B7-H2 Fc Chimera Alexa Fluor™ 488 (Catalog # AFG165) that produces 50% of the binding response is 20.0-200 ng/mL.
Specifications
Specifications
| Accession Number | O75144.2 |
| For Use With (Application) | Bioactivity |
| Formulation | Supplied as a 0.2 μm filtered solution in PBS with BSA as a carrier protein. |
| Gene ID (Entrez) | 23308 |
| Molecular Weight (g/mol) | M.W.-observed: 80-90 kDa, under reducing conditions., M.W.-theroretical: 53.4 kDa (monomer) |
| Quantity | 20 μg |
| Source | Mouse myeloma cell line, NS0-derived human B7-H2 protein Human B7-H2 (Asp19-Ser258) (C-terminus) Accession # O75144.2 DIEGRMD Human IgG1 (Pro100-Lys330) (N-terminus) |
| Storage Requirements | Protect from light. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after opening. 3 months, -20°C to -70°C under sterile conditions after opening. |
| Endotoxin Concentration | <0.10 EU / 1 μg of the protein by the LAL method. |
| Gene Alias | B7H2, B7H2B7 homolog 2, B7-H2B7-related protein 1, B7RP-1, B7RP1B7-like protein Gl50, B7RP-1 LICOS, CD275, CD275 antigen, GL50, GL50transmembrane protein B7-H2 ICOS ligand, ICOSL, ICOS-L, ICOSLG, inducible T-cell co-stimulator ligand, KIAA0653ICOS ligand |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction