missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human B7-H3 (4Ig)/B7-H3b Fc Chimera Protein
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
Description
Measured by its ability to inhibit anti-CD3 antibody induced IL-2 or IFN-gamma secretion by human T cells. The ED50 for this effect is 2-20 μg/mL.
Specifications
Specifications
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS. |
| Gene ID (Entrez) | 80381 |
| Molecular Weight (g/mol) | M.W.-observed: 100-120 kDa, under reducing conditions, M.W.-theoretical: 73 kDa |
| Quantity | 50 μg |
| Source | Human embryonic kidney cell, HEK293-derived human B7-H3 protein Human B7-H3 (4 Ig)/B7-H3b (Leu29-Thr461) (N-terminus) Accession # NP_001019907.1 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.10 EU / 1 μg of the protein by the LAL method. |
| Gene Alias | B7H3, B7H34Ig-B7-H3, B7-H3B7 homolog 3, CD276, CD276 antigen, CD276 molecule, Costimulatory molecule |
| Conjugate | Unconjugated |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction