missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human B7-H4 Fc Chimera Alexa Fluor™ 488 Protein
Click to view available options
:
20 μg
Description
Measured by flow cytometry for its ability to bind anti-human B7-H4 Antibody conjugated beads. The concentration of Recombinant Human B7-H4 Fc Chimera Alexa Fluor™ 488 (Catalog # AFG8870) that produces 50% of the binding response is 0.200-2.00 μg/mL.
Specifications
Specifications
| Accession Number | Q7Z7D3.1 |
| For Use With (Application) | Bioactivity |
| Formulation | Supplied as a 0.2 μm filtered solution in PBS with BSA as a carrier protein. |
| Molecular Weight (g/mol) | MolecularWeight-observed: 78-90 kDa, under reducing conditions., MolecularWeight-theroretical: 52 kDa |
| Quantity | 20 μg |
| Source | Human embryonic kidney cell, HEK293-derived human B7-H4 protein Human B7-H4 (Phe29-Ala258) (N-terminus) Accession # Q7Z7D3.1 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus) |
| Storage Requirements | Protect from light. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after opening. 3 months, -20°C to -70°C under sterile conditions after opening. |
| Endotoxin Concentration | <1.0 EU / 1 μg of the protein by the LAL method. |
| Conjugate | Alexa Fluor 488 |
| Purity or Quality Grade | >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction