missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human Bcl-2 Protein
Shop All Bio Techne Products
Click to view available options
Quantity:
100 μg
1000 μg
5 μg
500 μg
Unit Size:
1000µg
100µg
500µg
5µg
Description
Test in Process.
Specifications
Specifications
| Accession Number | P10415, P10415, P10415 |
| For Use With (Application) | PAGE |
| Formulation | 0.2 um filtered concentrated solution in 25 mM Tris-HCl, pH 8.0, 100mM NaCl, 1 mM DTT, Tween-80. |
| Gene ID (Entrez) | 596 |
| Molecular Weight (g/mol) | 23.2 kDa |
| Quantity | 100 μg |
| Storage Requirements | Store at -20C. Avoid freeze-thaw cycles. |
| Endotoxin Concentration | Less than 0.1 EU/ug of Bcl-2 as determined by LAL method. |
| Gene Alias | apoptosis regulator Bcl-2, B-cell CLL/lymphoma 2, Bcl-2 |
| Gene Symbol | BCL2 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction