missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Bcl 6 Protein

Product Code. 15976268
Click to view available options
Quantity:
10 μg
2 μg
5 μg
Unit Size:
10µg
2µg
5µg
This item is not returnable. View return policy

Product Code. 15976268

Brand: enQuireBio™ QP1113910ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the Bcl 6 was constructed and used to recombinantly synthesize the protein.

Specifications

Gene ID (Entrez) 604
Name Bcl 6 Protein
Quantity 10 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Gene Symbol BCL6
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag GST
Sequence MASPADSCIQ FTRHASDVLL NLNRLRSRDI LTDVVIVVSR EQFRAHKTVL MACSGLFYSI FTDQLKCNLS VINLDPEINP EGFCILLDFM YTSRLNLREG NIMAVMATAM YLQMEHVVDT CRKFIKASEA EMVSAIKPPR EEFLNSRMLM PQDIMAYRGR EVVENNLPLR SAPGCESRAF APSLYSGLST PPASYSMYSH LPVSSLLFSD EEFRDVRMPV ANPFPKERAL PCDSARPVPG EYSRPTLEVS PNVCHSNIYS PKETIPEEAR SDMHYSVAEG LKPAAPSARN APYFPCDKAS KEEERPSSED EIALHFEPPN APLNRKGLVS PQSPQKSDCQ PNSPTESCSS KNACILQASG SPPAKSPTDP KACNWKKYKF IVLNSLNQNA KPEGPEQAEL GRLSPRAYTA PPACQPPMEP ENLDLQSPTK LSASGEDSTI PQASRLNNIV NRSMTGSPRS SSESHSPLYM HPPKCTSCGS QSPQHAEMCL HTAGPTFPEE MGETQSEYSD SSCENGAFFC NECDCRFSEE ASLKRHTLQT HSDKPYKCDR CQASFRYKGN LASHKTVHTG EKPYRCNICG AQFNRPANLK THTRIHSGEK PYKCETCGAR FVQVAHLRAH VLIHTGEKPY PCEICGTRFR HLQTLKSHLR IHTGEKPYHC EKCNLHFRHK SQLRLHLRQK HGAITNTKVQ YRVSATDLPP ELPKAC
Buffer Bcl-6 protein in 50mM Tris-HCl, pH 7.5 and 10mM L-glutathione (reduced).
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.