missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human BCL-W / BCL2L2 Protein

A DNA sequence encoding the Homo sapiens (Human) BCL-W / BCL2L2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7839-ec-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16031111

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001186768.1
BCL-W / BCL2L2 Protein
Research Use Only
BCL2L2
Recombinant Protein
E. coli
ATPASAPDTR ALVADFVGYK LRQKGYVCGA GPGEGPAADP LHQAMRAAGD EFETRFRRTF SDLAAQLHVT PGSAQQRFTQ VSDELFQGGP NWGRLVAFFV FGAALCAESV NKEMEPLVGQ VQEWMVAYLE TQLADWIHSS GGWAEFTALY GDGALEEARR LREGNWASVR TVLTGAVALG ALVTVGAFFA SK
Greater than 90% as determined by SDS-PAGE.
599
500 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers