missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Beta Defensin 103AB Protein
Recombinant Protein
Brand: enQuireBio™ QP5323-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_001075020.1 | |
Beta Defensin 103AB Protein | |
Research Use Only | |
DEFB103AB | |
Human | |
Untagged | |
Sterile filtered (0.22 um) solution containing 0.1% TFA |
414325 | |
1 mg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
GIINTLQKYY CRVRGGRCAV LSCLPKEEQI GKCSTRGRKC CRRKK | |
Greater than 95% as determined by SDS-PAGE. |