missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Beta-defensin 124 Protein
A DNA sequence encoding the Homo sapiens (Human) Beta-defensin 124, was expressed in the hosts and tags indicated.
€ 202.00 - € 2155.00
Specifications
Accession Number | NP_001032589.1 |
---|---|
Gene ID (Entrez) | 245937 |
Name | Beta-defensin 124 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16091371
|
enQuireBio™
QP7883-YE-100UG |
100 μg |
€ 645.00
100µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16031381
|
enQuireBio™
QP7883-YE-500UG |
500 μg |
€ 1740.00
500µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16001381
|
enQuireBio™
QP7883-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16021381
|
enQuireBio™
QP7883-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16041381
|
enQuireBio™
QP7883-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16011381
|
enQuireBio™
QP7883-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001032589.1 | |
Beta-defensin 124 Protein | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
245937 | |
Research Use Only | |
DEFB124 | |
Recombinant Protein | |
Yeast | |
EFKRCWKGQG ACQTYCTRQE TYMHLCPDAS LCCLSYALKP PPVPKHEYE | |
Greater than 90% as determined by SDS-PAGE. |