missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Beta-defensin 124 Protein

A DNA sequence encoding the Homo sapiens (Human) Beta-defensin 124, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7883-ye-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16031381

  • € 1740.00 / 500µg
Estimated Shipment: 18-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001032589.1
Beta-defensin 124 Protein
Research Use Only
DEFB124
Recombinant Protein
Yeast
EFKRCWKGQG ACQTYCTRQE TYMHLCPDAS LCCLSYALKP PPVPKHEYE
Greater than 90% as determined by SDS-PAGE.
245937
500 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers