missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Beta-defensin 130 Protein

A DNA sequence encoding the Homo sapiens (Human) Beta-defensin 130, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9781-ye-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16028711

  • € 2650.00 / 500µg
Estimated Shipment: 27-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001032893.1
Beta-defensin 130 Protein
Research Use Only
DEFB130
Recombinant Protein
Yeast
GVIPGQKQCI ALKGVCRDKL CSTLDDTIGI CNEGKKCCRR WWILEPYPTP VPKGKSP
Greater than 90% as determined by SDS-PAGE.
100133267
500 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers