missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human BLID Protein

A DNA sequence encoding the Homo sapiens (Human) BLID, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7821-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16031001

  • € 3405.00 / 1mg
Estimated Shipment: 17-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001001786.2
BLID Protein
Research Use Only
BLID
Recombinant Protein
E. coli
MVTLLPIEGQ EIHFFEILES ECVLYTGWIE RASGSSIYPE AKARLPLEAL LGSNKEPMLP KETVLSLKRY NLGSSAMKRN VPGHVLQRPS YLTRIQVTLL CNSSAEAL
Greater than 80% as determined by SDS-PAGE.
414899
1 mg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers