missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human BMP-2 Protein
Click to view available options
Quantity:
1 mg
10 μg
100 μg
2 μg
500 μg
Unit Size:
100µg
10µg
1mg
2µg
500µg
Specifications
Specifications
| Accession Number | NP_001191.1 |
| Gene ID (Entrez) | 650 |
| Name | BMP-2 Protein |
| Quantity | 100 μg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | Less than or equal to 1.00 EUs/ug as determined by the LAL method. |
| Gene Symbol | BMP2 |
| Biological Activity | E. coli synthesized BMP-2Shows bioactivity at a concentration of 250 ng/mL in the Alkaline phosphatase activity induced in ATDC-5 cells assay. The bioactivity acceptance criteria is set at 4 x 10^3 units/mg. The more potent HEK293 derived BMP2 has a specific activity of less than 40 ng/ml in ATDC-5 cell induced alk phos activity. |
| Product Type | Recombinant Protein |
| Cross Reactivity | Human |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction