missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human BMP-7 Biotinylated Protein

Product Code. 18762148
Click to view available options
Formulation:
Lyophilized from a 0.2μm filtered solution in HCl with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in HCl.
Unit Size:
10µg
This item is not returnable. View return policy

Product Code. 18762148

Brand: R&D Systems™ BT354B010

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Human BMP-7 Biotinylated Protein is derived from CHO. The Recombinant Human BMP-7 Biotinylated Protein has been validated for the following applications: Binding Activity.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P18075
Characterization Binding Activity
For Use With (Application) Binding Activity
Formulation Lyophilized from a 0.2μm filtered solution in HCl with BSA as a carrier protein.
Gene ID (Entrez) 655
Molecular Weight (g/mol) 16kDa (unlabeled)
Quantity 10 μg
Source Chinese Hamster Ovary cell line,CHO-derived human BMP-7 protein Ser293-His431
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.01EU per 1μg of the protein by the LAL method.
Gene Alias BMP7, BMP-7, bone morphogenetic protein 7, Eptotermin alfa, OP-1, OP-1OP1, Osteogenic protein 1
Product Type Recombinant Protein
Conjugate Biotin
Species Human
Reconstitution Reconstitute at 100μg/mL in 4 mM HCl.
Purity or Quality Grade >90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie Blue Staining.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.