missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human BPHL Protein
A DNA sequence encoding the Homo sapiens (Human) BPHL, was expressed in the hosts and tags indicated.
€ 160.00 - € 2385.00
Specifications
Accession Number | NP_001289706.1 |
---|---|
Gene ID (Entrez) | 670 |
Name | BPHL Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16090831
|
enQuireBio™
QP7794-EC-100UG |
100 μg |
€ 530.00
100µg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16010841
|
enQuireBio™
QP7794-EC-1MG |
1 mg |
€ 2385.00
1mg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16000841
|
enQuireBio™
QP7794-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16020841
|
enQuireBio™
QP7794-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16030841
|
enQuireBio™
QP7794-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16040841
|
enQuireBio™
QP7794-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001289706.1 | |
BPHL Protein | |
Not determined. | |
Full Length of Isoform 2 | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
670 | |
Research Use Only | |
BPHL | |
Recombinant Protein | |
E. coli | |
MPRNLLYSLL SSHLSPHFST SVTSAKVAVN GVQLHYQQTG EGDHAVLLLP GMLGSGETDF GPQLKNLNKK LFTVVAWDPR GYGHSRPPDR DFPADFFERD AKDAVDLMKA LKFKKVSLLG WSDGGITALI AAAKYPSYIH KMVIWGANAY VTDEDSMIYE GIRDVSKWSE RTRKPLEALY GYDYFARTCE KWVDGIRQFK HLPDGNICRH LLPRVQCPAL IVHGEKDPLV PRFHADFIHK HVKGSRLHLM PEGKHNLHLR FADEFNKLAE DFLQ | |
Greater than 90% as determined by SDS-PAGE. |