missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human BRCA1-associated Protein

A DNA sequence encoding the Homo sapiens (Human) BRCA1-associated protein, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9827-ye-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16028801

  • € 1740.00 / 500µg
Estimated Shipment: 21-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_006759.3
BRCA1-associated Protein
Research Use Only
BRAP
Recombinant Protein
Yeast
MSVSLVVIRL ELAEHSPVPA GFGFSAAAGE MSDEEIKKTT LASAVACLEG KSPGEKVAII HQHLGRREMT DVIIETMKSN PDELKTTVEE RKSSEASPTA QRSKDHSKEC INAAPDSPSK QLPDQISFFS GNPSVEIVHG IMHLYKTNKM TSLKEDVRRS AMLCILTVPA AMTSHDLMKF VAPFNEVIEQ MKIIRDSTPN QYMVLIKFRA QADADSFYMT CNGRQFNSIE DDVCQLVYVE RAEVLKSEDG ASLPVMDLTE LPKCTVCLER MDESVNGILT TLCNHSFHSQ CLQRWDDTTC PVCRYCQTPE PVEENKCFEC GVQENLWICL ICGHIGCGRY VSRHAYKHFE ETQHTYAMQL TNHRVWDYAG DNYVHRLVAS KTDGKIVQYE CEGDTCQEEK IDALQLEYSY LLTSQLESQR IYWENKIVRI EKDTAEEINN MKTKFKETIE KCDNLEHKLN DLLKEKQSVE RKCTQLNTKV AKLTNELKEE QEMNKCLRAN QVLLQNKLKE EERVLKETCD QKDLQITEIQ EQLRDVMFYL ETQQKINHLP AETRQEIQEG QINIAMASAS SPASSGGSGK LPSRKGRSKR GK
Greater than 90% as determined by SDS-PAGE.
8315
500 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers