missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human BRCA1-associated Protein
A DNA sequence encoding the Homo sapiens (Human) BRCA1-associated protein, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9827-ye-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_006759.3 | |
BRCA1-associated Protein | |
Research Use Only | |
BRAP | |
Recombinant Protein | |
Yeast | |
MSVSLVVIRL ELAEHSPVPA GFGFSAAAGE MSDEEIKKTT LASAVACLEG KSPGEKVAII HQHLGRREMT DVIIETMKSN PDELKTTVEE RKSSEASPTA QRSKDHSKEC INAAPDSPSK QLPDQISFFS GNPSVEIVHG IMHLYKTNKM TSLKEDVRRS AMLCILTVPA AMTSHDLMKF VAPFNEVIEQ MKIIRDSTPN QYMVLIKFRA QADADSFYMT CNGRQFNSIE DDVCQLVYVE RAEVLKSEDG ASLPVMDLTE LPKCTVCLER MDESVNGILT TLCNHSFHSQ CLQRWDDTTC PVCRYCQTPE PVEENKCFEC GVQENLWICL ICGHIGCGRY VSRHAYKHFE ETQHTYAMQL TNHRVWDYAG DNYVHRLVAS KTDGKIVQYE CEGDTCQEEK IDALQLEYSY LLTSQLESQR IYWENKIVRI EKDTAEEINN MKTKFKETIE KCDNLEHKLN DLLKEKQSVE RKCTQLNTKV AKLTNELKEE QEMNKCLRAN QVLLQNKLKE EERVLKETCD QKDLQITEIQ EQLRDVMFYL ETQQKINHLP AETRQEIQEG QINIAMASAS SPASSGGSGK LPSRKGRSKR GK | |
Greater than 90% as determined by SDS-PAGE. |
8315 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |