missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human C6ORF108 Protein

A cDNA sequence encoding the C6ORF108 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11241-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15936588

  • € 3595.00 / 1mg

Estimated Shipment: 06-02-2023
to see stock.
Add to basket

Description

Description

Specifications

Specifications

C6ORF108 Protein
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The C6ORF108 protein solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8), 1mM DTT, and 10% glycerol.
Greater than 95.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MAAAMVPGRS ESWERGEPGR PALYFCGSIR GGREDRTLYE RIVSRLRRFG TVLTEHVAAA ELGARGEEAAGGDRLIHEQD LEWLQQADVV VAEVTQPSLG VGYELGRAVA FNKRILCLFR PQSGRVLSAM IRGAADGSRF QVWDYEEGEV EALLDRYFEA DPPGQVAASP DPTT
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers