missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CADM1 Protein
A cDNA sequence encoding the CADM1 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11254-10ug
Additional Details : Weight : 0.01000kg
Specifications
23705 | |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSQNLFTKD VTVIEGEVAT ISCQVNKSDD SVIQLLNPNR QTIYFRDFRP LKDSRFQLLN FSSSELKVSL TNVSISDEGR YFCQLYTDPP QESYTTITVL VPPRNLMIDI QKDTAVEGEE IEVNCTAMAS KPATTIRWFK GNTELKGKSE VEEWSDMYTV TSQLMLKVHK EDDGVPVICQ VEHPAVTGNL QTQRYLEVQY KPQVHIQMTY PLQGLTREGD ALELTCEAIG KPQPVMVTWV RVDDEMPQHA VLSGPNLFIN NLNKTDNGTY RCEASNIVGK AHSDYMLYVY DPPTTIPPPT TTTTTTTTTT TTILTIITDS RAGEEGSIRA VDH | |
Greater than 85.0% as determined by SDS-PAGE. |
CADM1 Protein | |
Research Use Only | |
CADM1 | |
Human | |
His | |
CADM1 protein solution (0.25 mg/ml) containing Phosphate buffered saline (pH 7.4), 10% glycerol and 1mM DTT. |