missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CALR3 Protein
Specifications
Specifications
| Name | CALR3 Protein |
| Quantity | 1 mg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
| Product Type | Recombinant Protein |
| Cross Reactivity | Human |
| Species | E. coli |
| Protein Tag | Untagged |
| Sequence | MTVYFQEEFL DGEHWRNRWL QSTNDSRFGH FRLSSGKFYG HKEKDKGLQT TQNGRFYAIS ARFKPFSNKG KTLVIQYTVK HEQKMDCGGG YIKVFPADID QKNLNGKSQY YIMFGPDICG FDIKKVHVIL HFKNKYHENK KLIRCKVDGF THLYTLILRP DLSYDVKIDG QSIESGSIEY DWNLTSLKKE TSPAESKDWE QTKDNKAQDW EKHFLDASTS KQSDWNGDLD GDWPAPMLQK PPYQDGLKPE GIHKDVWLHR KMKNTDYLTQ YDLSEFENIG AIGLELWQVR SGTIFDNFLI TDDEEYADNF GKATWGETKG PEREMDAIQA KEEMKKAREE EEEELLSGKI NRHEHYFNQF HRRNEL |
| Buffer | CALR3 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.4M Urea, and 10% glycerol. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?