Learn More
enQuireBio™ Recombinant Human Carbonic Anhydrase II / CA2 Protein
A DNA sequence encoding the Homo sapiens (Human) Carbonic Anhydrase II / CA2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5743-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_000058.1 | |
Carbonic Anhydrase II / CA2 Protein | |
Research Use Only | |
CA2 | |
Recombinant Protein | |
E. coli | |
SHHWGYGKHN GPEHWHKDFP IAKGERQSPV DIDTHTAKYD PSLKPLSVSY DQATSLRILN NGHAFNVEFD DSQDKAVLKG GPLDGTYRLI QFHFHWGSLD GQGSEHTVDK KKYAAELHLV HWNTKYGDFG KAVQQPDGLA VLGIFLKVGS AKPGLQKVVD VLDSIKTKGK SADFTNFDPR GLLPESLDYW TYPGSLTTPP LLECVTWIVL KEPISVSSEQ VLKFRKLNFN GEGEPEELMV DNWRPAQPLK NRQIKASFK | |
Greater than 90% as determined by SDS-PAGE. |
760 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |