missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human Carbonic Anhydrase VI Protein
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
Description
The Recombinant Human Carbonic Anhydrase VI Protein is derived from NS0. The Recombinant Human Carbonic Anhydrase VI Protein has been validated for the following applications: Enzyme Activity.
Specifications
Specifications
| Formulation | Supplied as a 0.2μm filtered solution in Tris, NaCl, Brij and Glycerol. |
| Gene ID (Entrez) | 765 |
| Molecular Weight (g/mol) | 35 kDa |
| Name | Carbonic Anhydrase VI |
| Quantity | 10 μg |
| Source | Mouse myeloma cell line,NS0-derived human Carbonic Anhydrase VI protein Gln18-Asn308,with a C-terminal 10-His tag |
| Storage Requirements | Do not freeze. 3 months from date of receipt, 2 to 8° C as supplied. 1 month, 2 to 8° C under sterile conditions after opening. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Purity or Quality Grade | 90%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction