missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CAS1 domain-containing protein 1
A DNA sequence encoding the Homo sapiens (Human) CAS1 domain-containing protein 1, was expressed in the hosts and tags indicated.
€ 160.00 - € 2385.00
Specifications
Accession Number | NP_075051.4 |
---|---|
Gene ID (Entrez) | 64921 |
Name | CAS1 domain-containing protein 1 |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16091761
|
enQuireBio™
QP7946-EC-100UG |
100 μg |
€ 530.00
100µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16011771
|
enQuireBio™
QP7946-EC-1MG |
1 mg |
€ 2385.00
1mg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16001771
|
enQuireBio™
QP7946-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16021771
|
enQuireBio™
QP7946-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16031771
|
enQuireBio™
QP7946-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16041771
|
enQuireBio™
QP7946-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_075051.4 | |
CAS1 domain-containing protein 1 | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
64921 | |
Research Use Only | |
CASD1 | |
Recombinant Protein | |
E. coli | |
ASRRYRGNDS CEYLLSSGRF LGEKVWQPHS CMMHKYKISE AKNCLVDKHI AFIGDSRIRQ LFYSFVKIIN PQFKEEGNKH ENIPFEDKTA SVKVDFLWHP EVNGSMKQCI KVWTEDSIAK PHVIVAGAAT WSIKIHNGSS EALSQYKMNI TSIAPLLEKL AKTSDVYWVL QDPVYEDLLS ENRKMITNEK IDAYNEAAVS ILNSSTRNSK SNVKMFSVSK LIAQETIMES LDGLHLPESS RETTAMILMN VYCNKILKPV DGSCCQPRPP VTLIQ | |
Greater than 90% as determined by SDS-PAGE. |