missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Caspase-2 Protein

A DNA sequence encoding the Homo sapiens (Human) Caspase-2, was expressed in the hosts and tags indicated.

€ 160.00 - € 2385.00

Specifications

Accession Number NP_001215.1
Gene ID (Entrez) 835
Name Caspase-2 Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
View More Specs

Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15917569
View Documents Promotion Details
enQuireBio™
QP5762-EC-100UG
100 μg
€ 530.00
100µg
View alternative products
Save Up to 
»
Estimated Shipment: 28-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15937569
View Documents Promotion Details
enQuireBio™
QP5762-EC-1MG
1 mg
€ 2385.00
1mg
View alternative products
Save Up to 
»
Estimated Shipment: 28-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15927569
View Documents Promotion Details
enQuireBio™
QP5762-EC-10UG
10 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
15947569
View Documents Promotion Details
enQuireBio™
QP5762-EC-200UG
200 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
15957569
View Documents Promotion Details
enQuireBio™
QP5762-EC-500UG
500 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
15967569
View Documents Promotion Details
enQuireBio™
QP5762-EC-50UG
50 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

Specifications

Specifications

NP_001215.1
Caspase-2 Protein
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
835
Research Use Only
CASP2
Recombinant Protein
E. coli
AAPSAGSWST FQHKELMAAD RGRRILGVCG MHPHHQETLK KNRVVLAKQL LLSELLEHLL EKDIITLEMR ELIQAKVGSF SQNVELLNLL PKRGPQAFDA FCEALRETKQ GHLEDMLLTT LSGLQHVLPP LSCDYDLSLP FPVCESCPLY KKLRLSTDTV EHSLDNKDGP VCLQVKPCTP EFYQTHFQLA YRLQSRPRGL ALVLSNVHFT GEKELEFRSG GDVDHSTLVT LFKLLGYDVH VLCDQTAQEM QEKLQNFAQL PAHRVTDSCI VALLSHGVEG AIYGVDGKLL QLQEVFQLFD NANCPSLQNK PKMFFIQACR GDETDRGVDQ QDGKNHAGSP GCEESDAGKE KLPKMRLPTR SDMICGYACL KGTAAMRNTK RGSWYIEALA QVFSERACDM HVADMLVKVN ALIKDREGYA PGTEFHRCKE MSEYCSTLCR HLYLFPGHPP T
Greater than 90% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications