missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Caspase-5 Protein
A DNA sequence encoding the Homo sapiens (Human) Caspase-5, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5763-ye-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001129581.1 | |
Caspase-5 Protein | |
Research Use Only | |
CASP5 | |
Recombinant Protein | |
Yeast | |
DQKITSVKPL LQIEAGPPES AESTNILKLC PREEFLRLCK KNHDEIYPIK KREDRRRLAL IICNTKFDHL PARNGAHYDI VGMKRLLQGL GYTVVDEKNL TARDMESVLR AFAARPEHKS SDSTFLVLMS HGILEGICGT AHKKKKPDVL LYDTIFQIFN NRNCLSLKDK PKVIIVQACR GEKHGELWVR D | |
Greater than 90% as determined by SDS-PAGE. |
838 | |
500 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |