missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human CCL3L1/LD78 beta Protein
Shop All Bio Techne Products
Click to view available options
Quantity:
1 mg
100 μg
5 μg
500 μg
Unit Size:
100µg
1mg
500µg
5µg
Description
CCL3L1 Protein is fully biologically active when compared to standard. The biological activity determined by a chemotaxis bioassay using human monocytes is in a concentration range of 1.0-10 ng/mL.
Specifications
Specifications
| Accession Number | P16619 |
| Concentration | LYOPH |
| For Use With (Application) | PAGE, Bioactivity |
| Formulation | Lyophilized from a 0.2 um filtered concentrated solution in PBS, pH 7.4. |
| Gene ID (Entrez) | 6349 |
| Molecular Weight (g/mol) | 7.8 kDa |
| Quantity | 100 μg |
| Storage Requirements | Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles. |
| Endotoxin Concentration | Less than 1 EU/ug of CCL3L1/LD78 beta as determined by LAL method. |
| Gene Alias | C-C motif chemokine 3-like 1, CCL3L3, chemokine (C-C motif) ligand 3-like 1, D17S1718, G0/G1 switch regulatory protein 19-2, G0S19-2LD78, LD78BETA, LD78-beta(1-70), MGC104178, MGC12815, MGC182017, MIP1AP, PAT 464.2, SCYA3L, SCYA3L1464.2, small inducible cytokine A3-like 1, Small-inducible cytokine A3-like 1, Tonsillar lymphocyte LD78 beta protein |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction