missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CCL5 / RANTES Protein

Recombinant Protein

Brand:  enQuireBio™ QP5261-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15935789

  • € 4240.00 / 1mg
Estimated Shipment: 12-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_002976.2
CCL5 / RANTES Protein
Research Use Only
CCL5
Recombinant Protein
E. coli
SPYSSDTTPC CFAYIARPLP RAHIKEYFYT SGKCSNPAVV FVTRKNRQVC ANPEKKWVRE YINSLEMS
Greater than 95% as determined by SDS-PAGE.
6352
1 mg
Less than or equal to 1.00 EUs/ug as determined by the LAL method.
Shows bioactivity in the THP-1 chemotaxis assay.
Human
Untagged
Sterile filtered (0.22 um) solution containing 0.1% TFA
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers