missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CCNA2 Protein
A cDNA sequence encoding the CCNA2 was constructed and used to recombinantly synthesize the protein.
€ 137.00 - € 4090.00
Specifications
Gene ID (Entrez) | 890 |
---|---|
Name | CCNA2 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gene Symbol | CCNA2 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15946748
|
enQuireBio™
QP11294-5UG |
5 μg |
€ 137.00
5µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15936748
|
enQuireBio™
QP11294-20UG |
20 μg |
€ 212.00
20µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15926748
|
enQuireBio™
QP11294-1MG |
1 mg |
€ 4090.00
1mg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
890 | |
Research Use Only | |
CCNA2 | |
Human | |
His | |
CCNA2 protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 20% glycerol, 0.15M NaCl and 1mM DTT. |
CCNA2 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMLGNSA PGPATREAGS ALLALQQTAL QEDQENINPE KAAPVQQPRT RAALAVLKSG NPRGLAQQQR PKTRRVAPLK DLPVNDEHVT VPPWKANSKQ PAFTIHVDEA EKEAQKKPAE SQKIEREDAL AFNSAISLPG PRKPLVPLDY PMDGSFESPHTMDMSIVLED EKPVSVNEVP DYHEDIHTYL REMEVKCKPK VGYMKKQPDI TNSMRAILVD WLVEVGEEYK LQNETLHLAV NYIDRFLSSM SVLRGKLQLV GTAAMLLASK FEEIYPPEVA EFVYITDDTY TKKQVLRMEH LVLKVLTFDL AAPTVNQFLT QYFLHQQPAN CKVESLAMFLGELSLIDADP YLKYLPSVIA GAAFHLALYT VTGQSWPESL IRKTGYTLES LKPCLMDLHQ TYLKAPQHAQ QSIREKYKNS KYHGVSLLNP PETLNL | |
Greater than 90% as determined by SDS-PAGE. |