missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CD300LF / LMIR3 Protein
A DNA sequence encoding the Homo sapiens (Human) CD300LF / LMIR3, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7829-ye-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001276011.1 | |
CD300LF / LMIR3 Protein | |
Research Use Only | |
CD300LF | |
Recombinant Protein | |
Yeast | |
TQITGPTTVN GLERGSLTVQ CVYRSGWETY LKWWCRGAIW RDCKILVKTS GSEQEVKRDR VSIKDNQKNR TFTVTMEDLM KTDADTYWCG IEKTGNDLGV TVQVTIDPAP VTQEETSSSP TLTGHHLDNR HKLLKLS | |
Greater than 90% as determined by SDS-PAGE. |
146722 | |
500 μg | |
Not determined. | |
Extracellular Domain | |
Human | |
His | |
Tris-based buffer, 50% glycerol |