missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human CD47 Fc Chimera Protein

Product Code. 18187420
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
This item is not returnable. View return policy

Product Code. 18187420

Brand: R&D Systems™ 4670CD050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Measured by its binding ability in a functional ELISA. When Recombinant Human (rh) SIRP alpha/CD172a Fc Chimera (Catalog # 4546-SA ) is coated at 2 μg/mL, Recombinant Human CD47 Fc Chimera binds with an apparent Kd <1 nM.

Optimal dilutions should be determined by each laboratory for each application.

Also measured by its ability to antagonize red blood cell adhesion to immobilized rhSIRP alpha Fc Chimera. The ED50 for this effect is 0.5-2.0 μg/mL.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q08722
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS.
Gene ID (Entrez) 961
Molecular Weight (g/mol) M.W. (Observed): 60-65 kDa, reducing conditions; M.W. (theoretical): 40.3 kDa (monomer)
Name CD47 Fc Chimera
Quantity 50 μg
Source Mouse myeloma cell line, NS0-derived human CD47 protein Human CD47 (Gln19-Pro139) (N-terminus) Accession # Q08722 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Structural Form Disulfide-linked homodimer
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Symbol CD47
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute at 200 μg/mL in PBS.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein CD47
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.