missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CD79B Protein
A cDNA sequence encoding the CD79B was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11348-HIS-20ug
Additional Details : Weight : 0.01000kg
Specifications
974 | |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMARSEDR YRNPKGSACS RIWQSPRFIA RKRGFTVKMH CYMNSASGNV SWLWKQEMDE NPQQLKLEKG RMEESQNESL ATLTIQGIRF EDNGIYFCQQ KCNNTSEVYQ GCGTELRVMG FSTLAQLKQR NTLKD | |
Greater than 90% as determined by SDS-PAGE. |
CD79B Protein | |
Research Use Only | |
CD79B | |
Human | |
His | |
CD79B protein solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 2M UREA and 10% glycerol. |